Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 4a Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002267R
Description
Cullin 4a Polyclonal antibody specifically detects Cullin 4a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Cullin 4a | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CUL-4A, cullin 4A, cullin-4A | |
The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 759) of human Cullin 4A using the numbering given in entry NP_001008895.1 (GeneID 8451). | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
8451 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction