Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 4a Antibody (GT885), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313594
This item is not returnable.
View return policy
Description
Cullin 4a Monoclonal antibody specifically detects Cullin 4a in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Cullin 4a | |
| Monoclonal | |
| Unconjugated | |
| CUL-4A, cullin 4A, cullin-4A | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Cullin 4a. The exact sequence is proprietary. | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 8451 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| GT885 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction