Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cullin 4A Recombinant Rabbit Monoclonal Antibody (5P4M1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535630
Description
Immunogen sequence: FKHKLFRIKI NQIQMKETVE EQVSTTERVF QDRQYQIDAA IVRIMKMRKT LGHNLLVSEL YNQLKFPVKP GDLKKRIESL IDRDYMERDK DNPNQYHYVA.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications
Cullin 4A | |
Recombinant Monoclonal | |
0.25 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13619 | |
CUL4A | |
A synthetic peptide corresponding to a sequence within amino acids 660-759 of human Cullin 4A (Q13619). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5P4M1 | |
Unconjugated | |
CUL4A | |
2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8451 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction