Learn More
Invitrogen™ Cullin 4A Monoclonal Antibody (GT2435)

Description
Positive Control: HeLa Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Cullin 4A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT2435 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CUL4A |
| Gene Accession No. | Q13619 |
| Gene Alias | 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.