Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cullin 4A Monoclonal Antibody (GT885)
GREENER_CHOICE

Catalog No. PIMA527796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527796 Supplier Invitrogen™ Supplier No. MA527796
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: HeLa Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 4A
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT885
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CUL4A
Gene Accession No. Q13619
Gene Alias 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853
Gene Symbols CUL4A
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Human Cullin 4a. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.