Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cullin 4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585573
Description
Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, A375. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications
Cullin 4A | |
Polyclonal | |
Unconjugated | |
CUL4A | |
2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q13619 | |
CUL4A | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Cullin 4a. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction