Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cullin 4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596216 Supplier Invitrogen™ Supplier No. PA596216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, BT-474, HeLa, Mouse testis Immunogen sequence: MADEAPRKGS FSALVGRTNG LTKPAALAAA PAKPGGAGGS KKLVIKNFRD RPRLPDNYTQ DTWRKLHEAV RAVQSSTSIR YNLEELYQAV ENLCSHKVSP MLYKQLRQAC EDHVQAQILP FREDSLDSVL FLKKINTCWQ DHCRQMIMIR SIFLFLDRTY VLQNSTLPSI WDMGLELFRT HIISDKMVQS KTIDGILLLI ERERSGEAVD RSLLRSLLGM LSDLQVYKDS FELKFLEETN CLYAAEGQRL MQEREVPEYL NHVSKRLEEE GDRVITYLDH STQKPLIACV EKQLLGEHLT.

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 4A
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CUL4A
Gene Accession No. Q13619, Q3TCH7
Gene Alias 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853
Gene Symbols CUL4A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-300 of human Cullin 4A (NP_0010088951).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361181, 8451, 99375
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.