Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cullin-4A, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1584MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1584MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1584MI Supplier MilliporeSigma™ Supplier No. ABS1584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cullin-4A (UniProt Q13619; also known as CUL-4A) is encoded by the CUL4A gene (Gene ID 8451) in human. CUL-4A belongs to the cullin family of hydrophobic proteins that functions as the scaffold component of E3 ubiquitin ligase complexes. CUL-4A interacts with DNA damage binding protein 1 (DDB1) and ring of cullins (ROC1) to assemble the E3 complex, where DDB1 acts a linker protein for CUL4 and recruits substrates to the CUL4-ROC1 catalytic core through interaction with a subset of WD40 proteins. At least 90 DDB1-binding WD40 (DWD) proteins (also known as DCAF for Ddb1- and Cul4-associated factors and CDW for CUL4 and DDB1-associated WD40 repeats) are known, suggesting a large number of potential substrates for CUL4-ROC1 ligase complexes. CUL4-ROC1 E3 ligase complexes play important roles in cell cycle, DNA replication, genomic stability, haematopoiesis, and spermatogenesis. CUL-4A is highly expressed in testis and spleen, while poor expression is found in lung, liver, thymus, small intestine, and muscular tissues. CUL4A gene amplification has been demonstrated in many kinds of tumors, including hepatocellular carcinomas (HCC), squamous cell carcinomas, and adrenocortical carcinoma. CUL4A silencing is reported to suppress the motility of HCC cells and reverse their epithelial-mesenchymal transition (EMT) tendency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin-4A
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CUL4A
Gene Accession No. Q13619
Host Species Rabbit
Immunogen Synthetic linear peptide corresponding to human Cullin-4A spliced isoform 1 N-terminal end sequence.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001008895
Test Specificity This polyclonal antibody was raised againt human Cullin-4A spliced isoform 1 N-terminal end sequence not present in spliced isoform 2. This polyclonal antibody detects both non-NEDDylated and NEDDylated Cullin-4A (McCall, C.M., et al. (2008). Mol. Cell. Biol. 28(18):5621-5633; Hu, J., et al. (2004). Nat. Cell Biol. 6(10):1003-1009).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.