Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cullin 4A Recombinant Rabbit Monoclonal Antibody (JU07-33)
GREENER_CHOICE

Catalog No. PIMA534615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534615 Supplier Invitrogen™ Supplier No. MA534615
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Hela, A431, HUVEC, human liver cance tissue, human breast cancer tissue, human colon cancer tissue, human kidney tissue, human cervix tissue.

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 4A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JU07-33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CUL4A
Gene Accession No. Q13619
Gene Alias 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853
Gene Symbols CUL4A
Host Species Rabbit
Immunogen Synthetic peptide within Human Cullin 4a aa 710-759.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8451
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.