Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 4B Polyclonal Antibody
Cullin 4B Polyclonal Antibody
Supplier: Thermo Scientific PA550647
Description
Cullin 4B Polyclonal Antibody for Western Blot

Specifications
Cullin 4B | |
Polyclonal | |
Unconjugated | |
CUL4B | |
2700050M05Rik; AA409770; CUL4B; CUL-4B; cullin 4B; cullin-4B; EGK_20888; KIAA0695; mKIAA0695; MRXHF2; MRXS15; MRXSC; RGD1564494; SFM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8450 | |
-20°C | |
Liquid |
Western Blot | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q13620 | |
CUL4B | |
Synthetic peptide corresponding to a region derived from internal residues of human cullin 4B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction