Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cullin 4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552864
Description
Immunogen sequence: GDGNDDEATT SKDGGFSSPS PSAAAAAQEV RSATDGNTST TPPTSAKKRK LNSSSSSSSN SSNEREDFDS TSSSSSTPPL QPRDSASPST SSFCLGVSVA ASSHVPIQKK LRFEDTLEFV GFDAKMAEES SSSSSSSSPT AATS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Core component of multiple cullin-RING-based E3 ubiquitin-protein ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of target proteins. The functional specificity of the E3 ubiquitin-protein ligase complex depends on the variable substrate recognition subunit. CUL4B may act within the complex as a scaffold protein, contributing to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. Plays a role as part of the E3 ubiquitin-protein ligase complex in polyubiquitination of CDT1, histone H2A, histone H3 and histone H4 in response to radiation-induced DNA damage. Targeted to UV damaged chromatin by DDB2 and may be important for DNA repair and DNA replication. Required for ubiquitination of cyclin E, and consequently, normal G1 cell cycle progression. Regulates the mammalian target-of-rapamycin (mTOR) pathway involved in control of cell growth, size and metabolism. Specific CUL4B regulation of the mTORC1-mediated pathway is dependent upon 26S proteasome function and requires interaction between CUL4B and MLST8.
Specifications
Cullin 4B | |
Polyclonal | |
Unconjugated | |
CUL4B | |
2700050M05Rik; AA409770; CUL4B; CUL-4B; cullin 4B; cullin-4B; EGK_20888; KIAA0695; mKIAA0695; MRXHF2; MRXS15; MRXSC; RGD1564494; SFM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
302502, 72584, 8450 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2A432, Q13620 | |
CUL4B | |
Recombinant protein corresponding to Human Cullin 4B. Recombinant protein control fragment (Product #RP-92221). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction