Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cullin 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102750 Supplier Invitrogen™ Supplier No. PA5102750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CUL5.

CUL5 is a core component of multiple SCF-like ECS (Elongin-Cullin 2/5-SOCS-box protein) E3 ubiquitin-protein ligase complexes, which mediate the ubiquitination and subsequent proteasomal degradation of target proteins. As a scaffold protein may contribute to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. The functional specificity of the E3 ubiquitin-protein ligase complex depends on the variable substrate recognition component. ECS(SOCS1) seems to direct ubiquitination of JAk2. It seems to be involved poteosomal degradation of p53/TP53 stimulated by adenovirus E1B-55 kDa protein and may form a cell surface vasopressin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CUL5
Gene Accession No. Q93034, Q9D5V5, Q9JJ31
Gene Alias 4921514I20Rik; 8430423K24Rik; AI852817; C030032G03Rik; C330021I08Rik; CUL5; CUL-5; cullin 5; cullin-5; Cullin-5 (vasopressin-activated calcium-mobilizing receptor-1); Vacm1; VACM-1; vasopressin-activated calcium-mobilizing receptor 1; vasopressin-activated calcium-mobilizing receptor protein; Vasopressin-activated calcium-mobilizing receptor-1
Gene Symbols CUL5
Host Species Rabbit
Immunogen A synthesized peptide derived from human CUL5(Accession Q93034), corresponding to amino acid residues A375-L425.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64624, 75717, 8065
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.