Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 5 Rabbit anti-Human, Mouse, Rat, Clone: S02-5K8, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Cullin 5 |
---|---|
Clone | S02-5K8 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
Classification | Monoclonal |
Description
Cullin 5 Monoclonal antibody specifically detects Cullin 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunocytochemistrySpecifications
Cullin 5 | |
Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CUL-5, cullin 5, VACM-1cullin-5, VACM1Cullin-5 (vasopressin-activated calcium-mobilizing receptor-1), Vasopressin-activated calcium-mobilizing receptor 1, Vasopressin-activated calcium-mobilizing receptor-1 | |
Recombinant protein of human Cullin 5 (Uniprot#: Q93034) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S02-5K8 | |
Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
8065 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title