Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin-7 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100602AF532
Description
Cullin-7 Polyclonal antibody specifically detects Cullin-7 in Human, Mouse samples. It is validated for Western BlotSpecifications
Cullin-7 | |
Polyclonal | |
Western Blot | |
CUL-7, cullin 7, cullin-7, KIAA0076dJ20C7.5 | |
A synthetic peptide, mapping to a region between residue 1650 and the C-terminus (residue 1698) of human Cullin homolog 7 using the numbering given in SwissProt entry Q14999 (GeneID 9820). | |
0.1 mL | |
Angiogenesis, DNA Repair | |
9820 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction