Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cullin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556337
Description
Immunogen sequence: FFIKKLDGPD HQEVLQILQE NLDGEILDDE ILAELAVPIE LAQDLLLTLP QRLNDSALRD LINCHVYKKY GPEALAGNQA YPSLLEAQED V Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 60%.
The protein encoded by this gene is a component of an E3 ubiquitin-protein ligase complex. The encoded protein interacts with TP53, CUL9, and FBXW8 proteins. Defects in this gene are a cause of 3M syndrome type 1 (3M1). Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Cullin 7 | |
Polyclonal | |
Unconjugated | |
CUL7 | |
2510004L20Rik; 3M1; AA409809; C230011P08Rik; CUL7; CUL-7; cullin 7; cullin-7; dJ20C7.5; KIAA0076; p185; p193 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14999 | |
CUL7 | |
Recombinant protein corresponding to Human Cullin 7. Recombinant protein control fragment (Product #RP-94604). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction