Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cullin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114724 Supplier Thermo Scientific Supplier No. PA5114724
Add to Cart
Add to Cart

Cullin 7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CUL7
Gene Accession No. Q14999, Q8VE73
Gene Alias 2510004L20Rik; 3M1; AA409809; C230011P08Rik; CUL7; CUL-7; cullin 7; cullin-7; dJ20C7.5; KIAA0076; p185; p193
Gene Symbols CUL7
Host Species Rabbit
Immunogen A synthesized peptide derived from human CUL7(Accession Q14999), corresponding to amino acid residues C1405-E1455.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66515, 9820
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.