Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Culti-Loops™ API™ NH QC Set

Catalog No. r4653070
Click to view available options
Quantity:
1/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ API NH Set contains quality control organisms supplied in Culti-Loops format.

Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Includes:

Haemophilus influenzae Type b ATCC™ 10211™†

Haemophilus paraphrophilus ATCC™ 49917™†

Neisseria gonorrhoeae ATCC™ 31426™†

Ordering Alerts:

Individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 10211TM*
Description API NH QC Set/3
Format Culti-Loops
Product Type Culti-Loops QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Packaging Type 1 Loop/Pack
Quantity 1/Pk.

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.