Learn More
Thermo Scientific™ Culti-Loops™ Coagulase Test QC Set
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Staphylococcus aureus subsp. aureus ATCC™ 25923™†
Staphylococcus epidermidis ATCC™ 12228™†
Ordering Information:
Individually sealed in a foil pouch and packaged five loops per package.

Specifications
Specifications
ATCC® | 25923TM* |
Certifications/Compliance | CLIA '88 Compliant. Culti-Loops are CE Marked. |
Description | CLIA '88 Compliance Coagulase Test QC Set/2 |
Format | Culti-Loops |
Product Type | QC set |
Target Organism Class | Organism Sets |
Test Type | Qualitative QC |
Quantity | 2 x 5 Loops |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.