Learn More
Thermo Scientific™ Culti-Loops™ Staphylococcus saprophyticus ATCC™ 35552™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
ATCC® | 35552TM* |
Certifications/Compliance | Culti-Loops are CE Marked. |
Description | Staphylococcus saprophyticus |
Product Type | Single Organism |
Target Organism Class | Cocci |
Test Type | Qualitative QC |
Packaging Type | 5 Loops/Pack |
Quantity | 5 Loops/Pk. |
Strain Designations | WDCM00008, API 1101 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.