Learn More
Thermo Scientific™ CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
Catalog No. 01182012
Change view
Click to view available options
No. of Shelves:
3
3 (Reinforced)
5 (Reinforced)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Shelves
01-182-012 5 (Reinforced)
01-257-037 3
01-182-013 3 (Reinforced)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-182-012 Supplier Thermo Scientific™ Supplier No. 50166268
Add to Cart
Add to Cart
This item is not returnable. View return policy
CultiMaxx™ Shelving System to support Nunc™ Cell Factory™ systems
These shelving systems are designed to support scale-out of the Nunc™ Cell Factory™ System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
These shelving systems are designed to support scale-out of the Nunc Cell Factory System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
Easy Installation
  • Shelving systems are retrofittable and can be self-installed.
TRUSTED_SUSTAINABILITY
Specifications
Description Shelving system to support up to 10 Nunc™ Cell Factory™ system with 4 layers for Heracell™ VIOS™ 250i (CR), Forma™ Steri-Cycle™ i250 (CR) CO2 Incubator
For Use With (Equipment) Heracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2 Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series
Material Stainless Steel
No. of Shelves 5 (Reinforced)
Type Shelving System
Includes 5 shelves with reinforced design and staggered to allow filter clearance, shelf supports and shelf rails