Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System

Catalog No. 01257035
Click to view available options
No. of Shelves:
10
3
3 (Reinforced)
5
5 (Reinforced)
6
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Catalog No. 01-257-035

Supplier: Thermo Scientific™ 332354G01

Only null left
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

CultiMaxx™ Shelving System to support Nunc™ Cell Factory™ systems

These shelving systems are designed to support scale-out of the Nunc™ Cell Factory™ System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.

These shelving systems are designed to support scale-out of the Nunc Cell Factory System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.

Easy Installation

  • Shelving systems are retrofittable and can be self-installed.
TRUSTED_SUSTAINABILITY

Specifications

Description Shelving system to support Nunc™ Cell Factory™ with 4 layers for Reach-In 821L CO2 Incubator
For Use With (Equipment) Reach-In 821L CO2 Incubator
Material Stainless Steel
No. of Shelves 10
Type Shelving System
Includes 10 levels of shelving
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.