Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex Antibiotic-free Mouse Laminin I, Pathclear
SDP

Catalog No. 340101002 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-010-1002 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-010-1002 Supplier R&D Systems™ Supplier No. 340101002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supports the attachment of MG63 osteosarcoma cells at <20 μg/mL.

Sterility Testing:

  • PathClear™ - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses.
  • No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP sterility testing guidelines.
  • Endotoxin concentration < 20 EU/mL by LAL assay.

Specifications

Product Line R&D Systems
Quantity 1 mg
Synonym LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.