Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Click to view available options
Quantity:
1 mL
10 mL (2 x 5 mL)
5 mL
Specifications
Specifications
| Quantity | 1 mL |
| Species | Mouse |
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction