Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumene, 99%

Catalog No. aaa11864ak
Click to view available options
Quantity:
250 mL
500 mL
2500 mL

CAS: 98-82-8 | C9H12 | 120.195 g/mol

Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

Solubility
Soluble in alcohol, ether, acetone, benzene, carbon tetrachloride. Insoluble in water.

Notes
Store at 2-8°C. Avoid prolonged storage with exposure to air. Incompatible with acids, oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96°C
Density 0.864
Boiling Point 152°C to 154°C
Flash Point 46°C (114°F)
Assay Percent Range 99%
Refractive Index 1.491
Quantity 250 mL
UN Number UN1918
Beilstein 1236613
Merck Index 14,2617
Solubility Information Soluble in alcohol,ether,acetone,benzene,carbon tetrachloride. Insoluble in water.
Formula Weight 120.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cumene
Show More Show Less
Hazard Category H226-H304-H335-H350
Hazard Statement GHS H Statement
H304-H226-H302-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPYLBENZENE
EINECSNumber 202-704-5
RTECSNumber GR8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.