Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumene, 99.9%, extra pure

Catalog No. p-94472
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 98-82-8 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.8620g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99.85% min. (GC)
Packaging Glass bottle
Refractive Index 1.4905 to 1.4925
Linear Formula C6H5CH(CH3)2
Quantity 5 mL
Beilstein 05,393
Specific Gravity 0.862
Merck Index 15,2607
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone
Viscosity 0.79 mPa.s (20°C)
Formula Weight 120.19
Percent Purity 99.9%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Cumene, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortabl
EINECSNumber 202-704-5
RTECSNumber GR8575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.