Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumyl hydroperoxide, 80%

Catalog No. AC349960010
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 80-15-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-15-9, 98-82-8, 617-94-7, 98-86-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
Synonym cumene hydroperoxide, cumyl hydroperoxide, hydroperoxide, 1-methyl-1-phenylethyl, cumenyl hydroperoxide, alpha,alpha-dimethylbenzyl hydroperoxide, cumolhydroperoxid, 7-cumyl hydroperoxide, cumolhydroperoxide, cument hydroperoxide, hydroperoxyde de cumene
PubChem CID 6629
ChEBI CHEBI:78673
SMILES CC(C)(OO)C1=CC=CC=C1

Specifications

Melting Point -30.0°C
Density 1.0600g/mL
Flash Point 83°C
Assay Percent Range 80 to 85% (Iodometry)
Packaging Plastic bottle
Linear Formula C6H5C(CH3)2OOH
Quantity 1 kg
Specific Gravity 1.06
Solubility Information Solubility in water: miscible.
IUPAC Name 2-hydroperoxypropan-2-ylbenzene
Formula Weight 152.19
Percent Purity 80%
Chemical Name or Material Cumyl hydroperoxide, 80%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Heating may cause a fire.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause re
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
EINECSNumber 201-254-7
RTECSNumber MX2450000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.