Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUP1-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144486 Supplier Invitrogen™ Supplier No. PA5144486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CUP1-1 Polyclonal Antibody for Western Blot, ELISA

Protects the cell against copper toxicity by tightly chelating copper ions. May also act as a depository for copper designated for the effective transfer into the apo forms of copper proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUP1-1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CUP1-1
Gene Accession No. P0CX80
Gene Alias MTH1
Gene Symbols CUP1-1
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) MTCU1 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856450
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.