Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupferron

Catalog No. p-94502
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-20-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00078422
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
PubChem CID 2724103
IUPAC Name azanium;N-oxido-N-phenylnitrous amide
SMILES N.ON(N=O)C1=CC=CC=C1

Specifications

Melting Point 150.0°C to 155.0°C
Ignition Residue 0.05% max
Linear Formula C6H4N(ONH4)NO
Quantity 25 g
Beilstein 16,I,395
Merck Index 15,2612
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether
Formula Weight 155.16
Grade ACS Reagent
Chemical Name or Material Cupferron
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Toxic if swallowed.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glo
EINECSNumber 205-183-2
RTECSNumber NC4725000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.