Learn More
Cupric Chloride Dihydrate, Purified (Copper (II) Chloride Dihydrate), Reagents
Specifications
Specifications
| Boiling Point | 993°C |
| Melting Point | 620°C |
| Color | Blue to Blue-Green |
| pH | 3 |
| Physical Form | Solid |
| Quantity | 500 g |
| Grade | Pure |
| Packaging | Poly Bottle |
| Specific Gravity | 2.54 |
| Chemical Name or Material | Cupric Chloride Dihydrate, Purified (Copper (II) Chloride Dihydrate) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.