Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Nitrate, Hydrate, Crystal, Purified, Spectrum™ Chemical
SDP

Catalog No. 18601909 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

C1407, 19004-19-4, Cu(NO3)2·2.5H2O

Spectrum™ Chemical Cupric Nitrate, Hydrate, Crystal, Purified is a also known as a copper nitrate and can be obtained as a crystalline solid as well as a hydrated form. Mostly used to produce copper oxide, it is also used as a polisher for other metals and in pyrotechnics.

Chemical Identifiers

CAS 19004-19-4
Molecular Formula Cu2H10N4O17
Molecular Weight (g/mol) 465.18
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
IUPAC Name dicopper(2+) pentahydrate tetranitrate
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 100%
UN Number UN1477
Quantity 2.5 kg
Grade Purified
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.