Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cupric Sulfate TS, 12.5% (w/v), Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Blue |
CAS Min % | 87.0, 10.9 |
CAS Max % | 91.0, 11.34 |
Percent Purity | 12.5% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
pH | ∼4 |
Quantity | 500 mL |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction