Learn More
Cupriethylenediamine, 1 Molar, Reagents
Cupriethylenediamine, 1 Molar, Reagent, for general laboratory use. Store under inert gas such as nitrogen or argon. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C21918504C1
| Quantity | 4 L |
|---|---|
| Packaging | Safety Coated Glass |
Chemical Identifiers
| 7732-18-5 , 13426-91-0 | |
| 18.02 | |
| CUED | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Dark Blue | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Safety Coated Glass | |
| 4 | |
| Non-sterile | |
| Cupriethylenediamine, 1 Molar |
| 7732-18-5 , 13426-91-0 | |
| CUED | |
| O | |
| 18.02 | |
| 1.1 | |
| 4 L | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.