Learn More
Cupriethylenediamine, 1 Molar, Reagents
Cupriethylenediamine, 1 Molar, Reagent, for general laboratory use. Store under inert gas such as nitrogen or argon. Manufactured in ISO 9001 facility.
$413.00 - $1824.00
Chemical Identifiers
| CAS | 7732-18-5 , 13426-91-0 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | CUED |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
01-337-301
|
Reagents Holdings Llc
C219180020Y |
20 L | Poly Drum |
Each for $1,824.00
|
|
||||
|
01-337-300
|
Reagents Holdings Llc
C21918004C |
4 L | Glass Bottle |
Each for $413.00
|
|
||||
|
01-337-299
|
Reagents Holdings Llc
C21918004QP |
4 L | Opaque HDPE |
Each for $454.00
|
|
||||
|
01-337-298
|
Reagents Holdings Llc
C21918504C1 |
4 L | Safety Coated Glass |
Each for $483.00
|
|
||||
Specifications
| Dark Blue | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 1.1 | |
| Non-sterile | |
| Cupriethylenediamine, 1 Molar |
| 7732-18-5 , 13426-91-0 | |
| CUED | |
| O | |
| 18.02 | |
| 4 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.