Learn More
Cupriethylenediamine, 1 Molar, Reagents
Cupriethylenediamine, 1 Molar, Reagent, for general laboratory use. Store under inert gas such as nitrogen or argon. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C21918004QP
| Quantity | 4 L |
|---|---|
| Packaging | Opaque HDPE |
Chemical Identifiers
| 7732-18-5 , 13426-91-0 | |
| 18.02 | |
| CUED | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Dark Blue | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Opaque HDPE | |
| 4 | |
| Non-sterile | |
| Cupriethylenediamine, 1 Molar |
| 7732-18-5 , 13426-91-0 | |
| CUED | |
| O | |
| 18.02 | |
| 1.1 | |
| 4 L | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.