Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Current Technologies Clear Acrylic Holders for HYPE-WIPE™ Towelettes

Catalog No. p-2669938
Click to view available options
Dimensions (L x W x H):
4 x 4 x 5.5 in.
4 x 7.75 x 3.5 in.
For Use With (Equipment):
HYPE-WIPE, 4 x 4 x 5.5 in. Bio-Hazard Wipes
Mini HYPE-WIPES
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce clutter on workbench, counters, trays, carts with clear acrylic dispensers for use with HYPE-WIPE™ and Mini HYPE-WIPE™ bleach towelettes

Specifications

Product Type Wiper Dispenser
Color Clear
Dimensions (L x W x H) 4 x 7.75 x 3.5 in.
Length (English) 4 in.
Length (Metric) 10.16 cm
Width (English) 7.75 in.
Width (Metric) 26.9 cm
Height (English) 3.5 in.
Height (Metric) 8.89 cm
Material Acrylic
For Use With (Application) Reduce clutter on workbench, counters, trays, carts
For Use With (Equipment) Mini HYPE-WIPES
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.