Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curriculum Mastery™ Flip Charts: Math
SCI_ED

Catalog No. p-3622969 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
1
2
3
5
6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S93933 3
S93932 2
S93936 6
S93935 5
S93931 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S93933 Supplier NewPath Learning Supplier No. 333001
Only null left
Add to Cart
Add to Cart

Includes

Activity guide with black-line copy masters, review questions, vocabulary terms, and suggested activities

Presents comprehensive review of science standards.

Curriculum Mastery Flip Charts provide a hands-on review of standards-based curriculum skills. Each developed to address the standards for its specific grade.

  • 10 Double-sided, laminated charts
  • Side 1 shows a colorful, graphic overview of the topic
  • Side 2 serves as a write-on/wipe-off activity chart featuring questions, labeling exercises, vocabulary review and more
  • Charts are mounted on sturdy easels for years of use
  • Chart Dimensions (L x W): 18 x 12 in. (45.7 x 30.5cm)

Topics include:
Grade 1 (S93931):

  • Numbers; Plane Shapes; Solid Figures; Time; Addition; Subtraction; Money; Fractions; Calendar; and Measuring
Grade 2 (S93932):
  • Addition Facts (two-digit); Subtraction Facts (two-digit); Hundred Counting Chart; Place Value to 100; Fractions; Telling Time; Length, Capacity, Weight, and Temperature; Data and Graphs; Ordinals; and Symmetry
Grade 3 (S93933):
  • Adding and Subtracting Three-Digit Numbers; Multiplication Concepts and Facts; Multiplication Table; Division Concepts and Facts; Counting Money; Addition and Subtraction Number Sense; All About Fractions; Decimals; Geometry and Measurement; and Problem-Solving Strategy
Grade 4 (S93934):
  • Place Value; Multiplying; Dividing; Adding and Subtracting Decimals; Polygons; Lines and Angles; Area, Volume, and Perimeter; Number Line; Adding and Subtracting Fractions; and Customary Units of Measurement
Grade 5 (S93935):
  • Data and Graphs; Multiplying and Dividing; Adding and Subtracting Fractions; Multiplying and Dividing Fractions; Understanding Percents; Order of Operations; Algebra; Probability; Congruent Shapes; and Angles
Grade 6 (S93936):
  • Commutative and Associative Properties; Decimal Operations; Graphing Ordered Pairs; Integers and Rational Numbers; Adding and Subtracting Integers; Multiplying and Dividing Integers; Writing Expressions; Solving Equations; Area of Triangles; and Area and Circumference of Circles

Specifications

Grade Level Elementary
Grade 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.