Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NoTrax™ Cushion Trax™ Anti-Fatigue Mat
GREENER_CHOICE

Catalog No. 19015791 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
5 x 3 ft.
12 x 3 ft.
75 x 3 ft.
75 x 4 ft.
75 x 5 ft.
Color:
Black Smoke
Black/Yellow Striped
Smoke Gray
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
19-015-791 12 x 3 ft. Black/Yellow Striped
19-015-794 5 x 3 ft. Black/Yellow Striped
19-015-795 5 x 3 ft. Smoke Gray
19-015-792 12 x 3 ft. Smoke Gray
19-015-793 12 x 3 ft. Black Smoke
19-015-802 75 x 3 ft. Smoke Gray
19-015-803 75 x 3 ft. Black Smoke
19-015-798 75 x 4 ft. Black/Yellow Striped
19-015-799 75 x 4 ft. Smoke Gray
19-015-800 75 x 4 ft. Black Smoke
19-015-797 75 x 5 ft. Black Smoke
19-015-796 5 x 3 ft. Black Smoke
19-015-801 75 x 3 ft. Black/Yellow Striped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-015-791 Supplier NoTrax™ Supplier No. 479S0312YB
Only null left
Add to Cart
Add to Cart

Cushion Trax™ is an anti-fatigue floor mat with a heavy-duty vinyl diamond-plate design and a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.

  • Cushion Trax™ is engineered to meet the toughest requirements of today's industrial applications
  • It is particularly suited for multi-shift operations in dry work areas where the worker is standing for prolonged periods of time such as workstations and assembly lines
  • The heavy-duty vinyl diamond-plate design provides non-directional traction, and is easy to clean
  • The top surface is combined with a dense closed cell foam base utilizing NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
  • NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate the slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
  • The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat

Specifications

Material Nitricell, PVC
Color Black/Yellow Striped
For Use With (Application) Manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual workstations. Suitable for commercial environments including hotels, reception desks, shop counters, department stores, banks, restaurants, etc.
Certifications/Compliance Certified by the NFSI (National Floor Safety Institute)
Type Anti-fatigue Floor Mat
Thickness (English) 9/16 in.
Length (Metric) 3.6576 m
Thickness (Metric) 14.2875 mm
Width (English) 3 ft.
Dimensions (L x W) 12 x 3 ft.
Length (English) 12 ft.
Width (Metric) 0.9144 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.