Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NoTrax™ Cushion Trax™ Anti-Fatigue Mat
Shop All NoTrax Products
Click to view available options
Dimensions (L x W):
5 x 3 ft.
12 x 3 ft.
75 x 3 ft.
75 x 4 ft.
75 x 5 ft.
Color:
Black Smoke
Black/Yellow Striped
Smoke Gray
Description
- Cushion Trax™ is engineered to meet the toughest requirements of today's industrial applications
- It is particularly suited for multi-shift operations in dry work areas where the worker is standing for prolonged periods of time such as workstations and assembly lines
- The heavy-duty vinyl diamond-plate design provides non-directional traction, and is easy to clean
- The top surface is combined with a dense closed cell foam base utilizing NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
- NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate the slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
- The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat
Specifications
Specifications
| Certifications/Compliance | Certified by the NFSI (National Floor Safety Institute) |
| Material | Nitricell, PVC |
| For Use With (Application) | Manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual workstations. Suitable for commercial environments including hotels, reception desks, shop counters, department stores, banks, restaurants, etc. |
| Color | Black/Yellow Striped |
| Type | Anti-fatigue Floor Mat |
| Dimensions (L x W) | 75 x 3 ft. |
| Thickness (English) | 9/16 in. |
| Length (English) | 75 ft. |
| Thickness (Metric) | 14.2875 mm |
| Length (Metric) | 22.86 m |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction