Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brew Plate™ Custom - 60mm plates of WLD
SCI_ED

Catalog No. s72044
Click to view available options
:
60mm, WLD

Brew Plate™ Custom - 60mm plates of WLD is part of a collection of prefilled, easy-to-use beer testing kits and bulk plates that detect microbes that spoil beer during the brewing process.

  • The Brew Plate Bulk Sleeves offer sleeves of 25 plates that allow the brewer to detect a wide scope of microbes known to cause contamination during the brewing process.
  • Each plate provides sections with various media to detect common wild yeast, aerobic (lives with oxygen) and anaerobic bacteria (lives without oxygen).
  • If contamination is detected, samples or plates can be sent to the Brew Plate Laboratory for microbial identification. Each bulk sleeve purchase comes with one free sequence identification.

Specifications

Content And Storage Refrigeration
Manufacturing Origin United States
Media Type Wallerstein Laboratories Differential Meda
Product Type Microbiology Media
For Use With (Application) Cultivating yeast and bacteria found in brewing.
Packaging Type Bulk Sleeve
Quantity 25 plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.