Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Custom Epoxy Work Surface

Catalog No. 06000386 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Epoxy
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-000-386 Epoxy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-000-386
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Custom Epoxy Work Surface is based on units of work surface designed and fabricated to suit your needs.

  • Contact your Fisher representative for details
  • 1 in. thick black epoxy resin

Order Info

Note: the price stated above ($ / Each) is per square foot.

Specifications

Product Type Work Surface
Includes Epoxy Work Surface
Warranty 2 Years
For Use With (Application) Work Surface
No. of Doors None
No. of Drawers None
Color Black
Height (English) 1 in.
Height (Metric) 2.5 cm
Material Epoxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.