Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ MGB Probe

Catalog No. A40005007
Encompass_Preferred
Change view
Click to view available options
5'Primer Modification:
ABY
Cyanine 5
FAM, NED, TET, VIC
JUN
Quantity:
20,000 pmol
20,000 pmol Scale
50,000 pmol
50,000 pmol Scale
6,000 pmol
6,000 pmol Scale
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. 5'Primer Modification Quantity
A40005007 Cyanine 5 20,000 pmol
A40005000 ABY 6,000 pmol
A40005001 ABY 20,000 pmol
A40005002 ABY 50,000 pmol
A40005003 JUN 6,000 pmol
A40005004 JUN 20,000 pmol
A40005005 JUN 50,000 pmol
A40005006 Cyanine 5 6,000 pmol
A40005008 Cyanine 5 50,000 pmol
43-160-34 FAM, NED, TET, VIC 6,000 pmol Scale
43-160-32 FAM, NED, TET, VIC 50,000 pmol Scale
43-160-33 FAM, NED, TET, VIC 20,000 pmol Scale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. A40005007 Supplier Applied Biosystems™ Supplier No. A40005007
Add to Cart
Add to Cart

Custom TaqMan probes are dual-labeled primers suitable for use in any real-time PCR application. They incorporate a 5' reporter dye and a 3' nonfluorescent quencher/minor groove binder (MGB). Choose among FAM, VIC, ABY, JUN, Cy5, TET, and NED as reporters.

Custom TaqMan probes are dual-labeled primers suitable for use in any real-time PCR application. They incorporate a 5' reporter dye and a 3' nonfluorescent quencher/Minor Groove Binder (MGB). Choose from FAM, VIC, ABY, JUN, Cyanine 5, TET, and NED as reporters. Probes are HPLC-purified.

Applied Biosystems TaqMan MGB (minor groove binder) probes are dual-labeled probes used for real-time PCR applications using TaqMan chemistry. TaqMan MGB Probes incorporate a 5' fluorescent reporter dye and a 3' nonfluorescent quencher (NFQ). The NFQ offers the advantage of lower background signal, which results in better precision in quantitation. Custom TaqMan MGB probes are HPLC-purified and available with FAM, VIC, ABY, JUN, Cyanine 5, TET, and NED reporter dyes.

Shorter, more specific probes

TaqMan MGB probes include a MGB moiety at the 3' end that increases the melting temperature (Tm) of the probe and stabilizes probe/target hybrids. This means that TaqMan MGB probes can be significantly shorter than traditional probes, providing better sequence discrimination and flexibility to accommodate more targets.

Nonfluorescent quencher (NFQ) maximizes sensitivity

TaqMan MGB probes incorporate an NFQ to absorb (quench) signal from the fluorescent dye label at the other end of the probe. The properties of the NFQ combined with the short length of the MGB probe result in lower background signal than with non-MGB NFQ probes. Lower background means increased sensitivity and precision in your data.

Multiplexing

The availability of MGB probes for up to five channels allows for higher-order multiplexing, enabling detection of more targets from the same samples.

Guaranteed yield

The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount.

Approximate ship time

4–10 days in North America, 5-10 days in Europe

We also offer:

  • Applied Biosystems custom primers and other types of TaqMan probes.
  • TaqMan MGB probes and primers available as predesigned and pre-formulated TaqMan Assays.
  • Commercial manufacturing products and services, including OEM and GMP oligo manufacturing.

Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs.

TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification MGB-NFQ
5'Primer Modification Cyanine 5
Detection Method TaqMan
Form Liquid
Format Tube
PCR Method RT-qPCR
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Green Features Sustainable packaging
Internal Probe Modification None
Label or Dye Cy5
Product Line TaqMan
Product Type TaqMan™ Probe
Purification Method HPLC
Purity or Quality Grade HPLC
Quantity 20,000 pmol
For Use With (Application) Real Time PCR (qPCR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.