Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brew Plate™ Custom - Whole plates of LCSM

Click to view available options
:
LCSM
Description
- The Brew Plate Bulk Sleeves offer sleeves of 25 plates that allow the brewer to detect a wide scope of microbes known to cause contamination during the brewing process.
- Each plate provides sections with various media to detect common wild yeast, aerobic (lives with oxygen) and anaerobic bacteria (lives without oxygen).
- If contamination is detected, samples or plates can be sent to the Brew Plate Laboratory for microbial identification. Each bulk sleeve purchase comes with one free sequence identification.
Specifications
Specifications
Content And Storage | Refrigeration |
Manufacturing Origin | United States |
Media Type | Lin's Cupric Sulfate Medium |
Product Type | Microbiology Media |
For Use With (Application) | Cultivating yeast and bacteria found in brewing. |
Packaging Type | Bulk Sleeve |
Quantity | 25 Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction