Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves

Catalog No. 17200353
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves feature soft and flexible TPR pads that provide an additional layer of protection against impacts to the back of the hand and fingers. Nitrile coating on the palm improves grip capabilities when handling wet and oily parts.

  • A blend of high performance and cooling fibers extends comfort in warmer work environments
  • Soft and flexible TPR pads provide an additional layer of protection against blows to the back of the hand and fingers
  • Reinforced thumb crotch for extra protection between the thumb and forefinger
  • An ANSI Cut Level A4 ensures a high level of cut protection against sharp objects
  • Padded, sandy nitrile coated palm increases protection and improves grip
  • Ideal for handling parts in wet and oily environments

Specifications

ANSI Abrasion Rating ANSI 4
Applications Industrial
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material HPPE
Product Type Anti-impact Gloves
Size X-Large
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 4
Resistant To Cuts, Abrasions, Impacts, Punctures
Series I2469
Tear Rating 4
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.