Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542620 Supplier Invitrogen™ Supplier No. PA542620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AFVTCPNEKV AKEIARAVVE KRLAACVNLI PQITSIYEWK GKIEEDSEVL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

May forms part of a complex of membrane proteins attached to acetylcholinesterase (AChE). Target protein may play a role in anchoring acetylcholinesterase in neuronal cell membranes; may be involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUTA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CUTA
Gene Accession No. O60888
Gene Alias 0610039D01Rik; 1810022E02Rik; 1810060C03Rik; 2700094G22Rik; acetylcholinesterase-associated protein; ACHAP; AI326454; Brain acetylcholinesterase putative membrane anchor; C6orf82; Cuta; cutA divalent cation tolerance homolog; cutA divalent cation tolerance homolog (E. coli); CutA1; DASS-97D12.5; divalent cation tolerant protein CUTA; MGC111154; protein CutA; Unknown (protein for MGC:140443)
Gene Symbols CUTA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CUTA.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51596
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.