Learn More
Invitrogen™ CUTA Polyclonal Antibody

Description
Immunogen sequence: TSIYEWKGKI EEDSEVLMMI KTQSSLVPAL TDFVRSVHPY EVAEVIALPV EQGNFPYLQW VRQVTESVSD SITVLP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Specifications
Specifications
| Antigen | CUTA |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CUTA |
| Gene Accession No. | O60888 |
| Gene Alias | 0610039D01Rik; 1810022E02Rik; 1810060C03Rik; 2700094G22Rik; acetylcholinesterase-associated protein; ACHAP; AI326454; Brain acetylcholinesterase putative membrane anchor; C6orf82; Cuta; cutA divalent cation tolerance homolog; cutA divalent cation tolerance homolog (E. coli); CutA1; DASS-97D12.5; divalent cation tolerant protein CUTA; MGC111154; protein CutA; Unknown (protein for MGC:140443) |
| Gene Symbols | CUTA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.