Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CUTA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109607
Description
Immunogen sequence: FVWMPALLPV ASRLLLLPRV LLTMASGSPP TQPSPASDSG SGYVPGSVSA AFVTCPNEKV AKEIARAVVE KRLAACVNLI PQITSIYEWK GKIEEDSEVL MMIKTQSSLV PALTDFVRSV HPYEVAEVIA LPVEQG.
May forms part of a complex of membrane proteins attached to acetylcholinesterase (AChE). Target protein may play a role in anchoring acetylcholinesterase in neuronal cell membranes; may be involved in signal transduction.
Specifications
CUTA | |
Polyclonal | |
Unconjugated | |
CUTA | |
0610039D01Rik; 1810022E02Rik; 1810060C03Rik; 2700094G22Rik; acetylcholinesterase-associated protein; ACHAP; AI326454; Brain acetylcholinesterase putative membrane anchor; C6orf82; Cuta; cutA divalent cation tolerance homolog; cutA divalent cation tolerance homolog (E. coli); CutA1; DASS-97D12.5; divalent cation tolerant protein CUTA; MGC111154; protein CutA; Unknown (protein for MGC:140443) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67675 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CQ89 | |
CUTA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-156 of human CUTA (NP_001014840.1). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction