Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUTA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8400 Supplier Bioss Supplier No. BS8248R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May forms part of a complex of membrane proteins attached to acetylcholinesterase (AChE). Target protein may play a role in anchoring acetylcholinesterase in neuronal cell membranes; may be involved in signal transduction.

Specifications

Antigen CUTA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CUTA
Gene Accession No. O60888
Gene Alias 0610039D01Rik; 1810022E02Rik; 1810060C03Rik; 2700094G22Rik; Acetylcholinesterase-associated protein; ACHAP; AI326454; BOS_21290; Brain acetylcholinesterase putative membrane anchor; C6orf82; CUTA; cutA divalent cation tolerance homolog; cutA divalent cation tolerance homolog (E. coli); CutA1; DASS-97D12.5; divalent cation tolerant protein CUTA; LOC100357492; MGC111154; Protein CutA
Gene Symbols CUTA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CUTA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51596
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.