Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eBioscience™

Catalog No. 14985737
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-985-737 2 mg
50-112-2886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-985-737 Supplier Invitrogen™ Supplier No. 14985737
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This HECA-452 monoclonal antibody reacts with human Cutaneous Lymphocyte Antigen (CLA). This cell surface glycoprotein is most highly expressed on T cells in the skin. Nevertheless, CLA can also be detected on some peripheral T and NK cells, as well as monocytes, neutrophils and CD1a+ dendritic cells. Studies have determined that this antibody recognizes sialofucosylated glycans and sialyl Lewis x (sLex). CLA, which binds E-selectin and endothelial cell-leukocyte adhesion molecule 1 (ELAM-1), is involved in T lymphocyte infiltration and adhesion at cutaneous sites of inflammation. The HECA-452 antibody has been reported to inhibit sLex binding to E-selectin; however, this property is still under debate in the literature. Applications Reported: This HECA-452 antibody has been reported for use in flow cytometric analysis, western blotting, functional assays, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This HECA-452 antibody has been tested by flow cytometric analysis on human PBMCs and by immunohistochemistry on FFPE human tissue. The HECA-452 antibody can be used at less than or equal to 0.125 μg/test for flow cytometry and at less than or equal to 10 μg/mL. The antibody performs optimally with low pH antigen retrieval but high pH antigen retrieval can also be used. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than...

This cell surface glycoprotein is most highly expressed on T cells in the skin. Nevertheless, CLA can also be detected on some peripheral T and NK cells, as well as monocytes, neutrophils and CD1a+ dendritic cells. Studies have determined that this antibody recognizes sialofucosylated glycans and sialyl Lewis x (sLex). CLA, which binds E-selectin and endothelial cell-leukocyte adhesion molecule 1 (ELAM-1), is involved in T lymphocyte infiltration and adhesion at cutaneous sites of inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cutaneous Lymphocyte Antigen (CLA)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HECA-452
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias CLA
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.