Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUTC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112306 Supplier Invitrogen™ Supplier No. PA5112306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CUTC Polyclonal Antibody for Western Blot, IHC (P), IP

Members of the CUT family of copper transporters are associated with copper homeostasis and are involved in the uptake, storage, delivery, and efflux of copper (Gupta et al., 1995 ; Li et al., 2005 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUTC
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CUTC
Gene Accession No. Q9NTM9
Gene Alias 2310039I18Rik; AI326282; CGI-32; Copper homeostasis protein cutC homolog; Cutc; cutC copper transporter; cutC copper transporter homolog; cutC copper transporter homolog (E.coli); RP11-483F11.3
Gene Symbols CUTC
Host Species Rabbit
Immunogen E. coli-derived Human CUTC fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.