Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CUX1/Protein CASP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598395
Description
CUX1/Protein CASP Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
CUX1/Protein CASP | |
Polyclonal | |
Unconjugated | |
CUX1 | |
CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; Cux1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; protein CASP; putative protein product of Nbla10317 | |
Rabbit | |
Protein G | |
RUO | |
116639, 13047, 1523 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.7 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P39880, P53565, P70403, Q13948 | |
CUX1 | |
Recombinant Human Protein CASP protein (50-210AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction